Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001029858.3 | NP_001025029.2 | 6 | 1016 | |
XM_005266865.4 | XP_005266922.1 | 6 | 1316 | ||
GenBank mRNA | AK128717.1 | - | 4 | 364 | |
AK295860.1 | - | 6 | 713 | ||
BC028615.1 | - | 6 | 1019 | 4817165 | |
BC036493.1 | - | 6 | 784 | ||
BC114925.1 | - | 7 | 890 | 40025808 | |
BX640761.1 | - | 3 | 277 |