Gene Transcripts

Gene Symbol
Cdca7
Entrez Gene ID
311742
Gene Name
cell division cycle associated 7
Gene Aliases
Chromosome Location
Chr. 3: 59153281 - 59163916 on Build Rnor_6.0
UniGene ID
Rn.161800
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025693.1 NP_001020864.1 8 1075
GenBank mRNA BC098690.1 - 8 1075

View Full Product Details