Gene Transcripts

Gene Symbol
Iqub
Entrez Gene ID
296936
Gene Name
IQ motif and ubiquitin domain containing
Gene Aliases
RGD1562359, Trs4
Chromosome Location
Chr. 4: 51482169 - 51586881 on Build Rnor_6.0
UniGene ID
Rn.11866
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034130.1 NP_001029302.1 7 1232
GenBank mRNA BC099086.1 - 7 1232
DQ132434.1 - 7 1279

View Full Product Details