Gene Transcripts
- Gene Symbol
- Apeg3
- Entrez Gene ID
- 619476
- Gene Name
- antisense paternally expressed gene 3
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Rn.49740
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB083820.1 |
- |
1 |
761 |
|
View Full Product Details