Gene Transcripts

Gene Symbol
C5orf15
Entrez Gene ID
56951
Gene Name
chromosome 5 open reading frame 15
Gene Aliases
HTGN29, KCT2
Chromosome Location
Chr. 5: 133955507 - 133968715 on Build GRCh38
UniGene ID
Hs.744034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020199.2 NP_064584.1 2 517
GenBank mRNA AF226055.1 - 2 590
AK074963.1 - 2 429
AK298515.1 - 3 326
AK315362.1 - 2 415
AY157581.1 - 2 426
BC020875.1 - 2 408 4610607
BP245947.1 - 2 517

View Full Product Details