Gene Transcripts

Gene Symbol
Tas2r126
Entrez Gene ID
387353
Gene Name
taste receptor, type 2, member 126
Gene Aliases
T2R12, T2R26, Tas2r26, Tas2r41, mGR26, mt2r35
Chromosome Location
Chr. 6: 42434535 - 42435461 on Build GRCm38
UniGene ID
Mm.377914
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207028.1 NP_996911.1 1 291
GenBank mRNA BC126974.1 - 1 291

View Full Product Details