Gene Transcripts

Gene Symbol
POGLUT1
Entrez Gene ID
56983
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
Chromosome Location
Chr. 3: 119468938 - 119494708 on Build GRCh38
UniGene ID
Hs.231750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152305.2 NP_689518.1 1, 2 177
NR_024265.1 1, 2 177
XM_017006879.1 XP_016862368.1 1, 2 123
XM_017006880.1 XP_016862369.1 1, 2 123
GenBank mRNA AF168711.1 - 1, 2 128
AK075444.1 - 1, 2 113
AK222937.1 - 1, 2 137
AK295984.1 - 1, 2 152
AK301002.1 - 1, 2 152
AK315367.1 - 1, 2 152
AY298903.1 - 1, 2 163
AY358581.1 - 1, 2 120
BC030614.2 - 1, 2 163 4796951
BC048810.1 - 1, 2 121 5243487
BP347179.1 - 1, 2 177

View Full Product Details