Gene Transcripts

Gene Symbol
Trcg1
Entrez Gene ID
541610
Gene Name
taste receptor cell gene 1
Gene Aliases
Chromosome Location
Chr. 9: 57236460 - 57252175 on Build GRCm38
UniGene ID
Mm.378036
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014398.2 NP_001014420.2 3 322
GenBank mRNA AY701870.1 - 3 322

View Full Product Details