Gene Transcripts
- Gene Symbol
- 2700094K13Rik
- Entrez Gene ID
- 72657
- Gene Name
- RIKEN cDNA 2700094K13 gene
- Gene Aliases
- Selh
- Chromosome Location
- Chr. 2: 84669221 - 84670708 on Build GRCm38
- UniGene ID
- Mm.259293
- Species
- Mus musculus
- Gene Symbol
- Lin54
- Entrez Gene ID
- 231506
- Gene Name
- lin-54 homolog (C. elegans)
- Gene Aliases
- 4832414L08, AI461788, mKIAA2037
- Chromosome Location
- Chr. 5: 100442033 - 100500649 on Build GRCm38
- UniGene ID
- Mm.212568
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC055819.1 |
- |
|
793 |
|
View Full Product Details