Gene Transcripts
- Gene Symbol
- SPEM1
- Entrez Gene ID
- 374768
- Gene Name
- spermatid maturation 1
- Gene Aliases
- C17orf83
- Chromosome Location
- Chr. 17: 7420360 - 7421632 on Build GRCh38
- UniGene ID
- Hs.710599
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097400.1 |
- |
2 |
1049 |
|
View Full Product Details