Gene Transcripts

Gene Symbol
C5orf56
Entrez Gene ID
441108
Gene Name
chromosome 5 open reading frame 56
Gene Aliases
Chromosome Location
Chr. 5: 132410773 - 132476044 on Build GRCh38
UniGene ID
Hs.658288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK096941.1 - 4 283
BC130299.1 - 3 181 40146330
BC130656.1 - 3 181 40146687

View Full Product Details