Gene Transcripts

Gene Symbol
TMEM9B
Entrez Gene ID
56674
Gene Name
TMEM9 domain family member B
Gene Aliases
C11orf15
Chromosome Location
Chr. 11: 8947201 - 8965011 on Build GRCh38
UniGene ID
Hs.501853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286094.1 NP_001273023.1 4 889
NM_001286095.1 NP_001273024.1 5 538
NM_020644.2 NP_065695.1 5 981
GenBank mRNA AB097017.1 - 5 606
AK074677.1 - 5 599
AK300712.1 - 5 461
AK313065.1 - 5 611
AK316216.1 - 4 520
AY359069.1 - 5 624
BC015884.1 - 4 516 3909465
BC040124.1 - 5 663 6053804
DC314550.1 - 5 538

View Full Product Details