Gene Transcripts

Gene Symbol
Oaf
Entrez Gene ID
315594
Gene Name
OAF homolog (Drosophila)
Gene Aliases
RGD1305890
Chromosome Location
Chr. 8: 47511718 - 47529689 on Build Rnor_6.0
UniGene ID
Rn.1638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014090.1 NP_001014112.1 2 502
GenBank mRNA BC079082.1 - 2 502

View Full Product Details