Gene Transcripts

Gene Symbol
VN1R1
Entrez Gene ID
57191
Gene Name
vomeronasal 1 receptor 1
Gene Aliases
V1RL1, VNR19I1, ZVNH1, ZVNR1
Chromosome Location
Chr. 19: 57455174 - 57456739 on Build GRCh38
UniGene ID
Hs.677423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020633.3 NP_065684.1 1 758
GenBank mRNA AF255342.1 - 1 505
AF302903.1 - 1 505
AK094452.1 - 1 1164
BC095508.1 - 1 548 6971941

View Full Product Details