Gene Transcripts

Gene Symbol
Zswim5
Entrez Gene ID
74464
Gene Name
zinc finger SWIM-type containing 5
Gene Aliases
4933426E21Rik, AI503093, mKIAA1511
Chromosome Location
Chr. 4: 116877402 - 116989105 on Build GRCm38
UniGene ID
Mm.218946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029912.2 NP_001025083.1 14 3187
GenBank mRNA AK122519.1 - 14 3187
BC089375.1 - 3 601

View Full Product Details