Gene Transcripts

Gene Symbol
Zfp27
Entrez Gene ID
22689
Gene Name
zinc finger protein 27
Gene Aliases
Zfp-27, mkr-4, mszf76
Chromosome Location
Chr. 7: 29893337 - 29906567 on Build GRCm38
UniGene ID
Mm.86747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037707.1 NP_001032796.1 5 1895
NM_001285797.1 NP_001272726.1 5 2167
NM_001285798.1 NP_001272727.1 5 2121
NM_011754.2 NP_035884.2 5 1890
GenBank mRNA AK028500.1 - 5 2122
AK135293.1 - 5 2171
AK147732.1 - 5 1885
BC059037.1 - 3 1834 6812360
M36515.1 - 2 767
X12593.1 - 2 767

View Full Product Details