Gene Transcripts

Gene Symbol
F8a
Entrez Gene ID
14070
Gene Name
factor 8-associated gene A
Gene Aliases
AI852759, DXUcsf1, F8a1
Chromosome Location
Chr. X: 73228306 - 73230795 on Build GRCm38
UniGene ID
Mm.165807
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007978.3 NP_032004.2 1 2062
GenBank mRNA AK033184.1 - 1 2065
AK046025.1 - 1 2078
AK046459.1 - 1 2077
AK054140.1 - 1 2063

View Full Product Details