Gene Transcripts

Gene Symbol
DHH
Entrez Gene ID
50846
Gene Name
desert hedgehog
Gene Aliases
GDXYM, HHG-3, SRXY7
Chromosome Location
Chr. 12: 49086656 - 49094819 on Build GRCh38
UniGene ID
Hs.524382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021044.3 NP_066382.1 1 579
GenBank mRNA AK313360.1 - 1 549
BC033507.1 - 1 579 5169286
HQ607678.1 - 1 583

View Full Product Details