Gene Transcripts

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
Chromosome Location
Chr. 20: 25407470 - 25448568 on Build GRCh38
UniGene ID
Hs.658464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021067.4 NP_066545.3 1, 2 224
NR_134574.1 1, 2 224
XM_017028162.1 XP_016883651.1 1, 2 181
GenBank mRNA AK294593.1 - 1, 2 178
BC012542.1 - 1, 2 225 4333095
BP290581.1 - 224
D80008.1 - 1, 2 180

View Full Product Details