Gene Transcripts

Gene Symbol
HNF1A
Entrez Gene ID
6927
Gene Name
HNF1 homeobox A
Gene Aliases
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
Chromosome Location
Chr. 12: 120978515 - 121002512 on Build GRCh38
UniGene ID
Hs.654455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000545.6 NP_000536.5 10 2330
NM_001306179.1 NP_001293108.1 10 2351
XM_005253931.3 XP_005253988.1 9 2476
GenBank mRNA HM116542.1 - 10 1946
HM116543.1 - 8 1735
HM116544.1 - 7 1389
HM116545.1 - 7 1259
HM116546.1 - 5 889
HM116549.1 - 4 589
HM116551.1 - 2 445
HM449088.1 - 9 2245
HM449089.1 - 9 1952
M57732.1 - 10 2152

View Full Product Details