Gene Transcripts
- Gene Symbol
- CRIP1
- Entrez Gene ID
- 1396
- Gene Name
- cysteine rich protein 1
- Gene Aliases
- CRHP, CRIP, CRP-1, CRP1
- Chromosome Location
- Chr. 14: 105486920 - 105488789 on Build GRCh38
- UniGene ID
- Hs.70327
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC002738.2 |
- |
|
406 |
|
BU500642.1 |
- |
|
416 |
|
View Full Product Details