Gene Transcripts
- Gene Symbol
- PTTG1
- Entrez Gene ID
- 9232
- Gene Name
- pituitary tumor-transforming 1
- Gene Aliases
- EAP1, HPTTG, PTTG, TUTR1
- Chromosome Location
- Chr. 5: 160421807 - 160428744 on Build GRCh38
- UniGene ID
- Hs.350966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282382.1 |
NP_001269311.1
|
4, 5 |
937 |
|
NM_001282383.1 |
NP_001269312.1
|
5, 6 |
791 |
|
NM_004219.3 |
NP_004210.1
|
5, 6 |
630 |
|
GenBank mRNA |
AF075242.1 |
- |
4, 5 |
554 |
|
AF095287.1 |
- |
5, 6 |
570 |
|
AJ223953.1 |
- |
4, 5 |
639 |
|
AK312209.1 |
- |
5, 6 |
608 |
|
BC101834.1 |
- |
5, 6 |
563 |
8069340 |
BC108894.1 |
- |
4, 5 |
534 |
|
BC108895.1 |
- |
4, 5 |
534 |
|
BC112071.1 |
- |
5, 6 |
563 |
8327539 |
BC113842.1 |
- |
4, 5 |
534 |
|
BC113896.1 |
- |
4, 5 |
534 |
|
BC128193.1 |
- |
4, 5 |
534 |
40113885 |
BC128194.1 |
- |
4, 5 |
534 |
40113886 |
CR457135.1 |
- |
4, 5 |
534 |
|
- Gene Symbol
- PTTG3P
- Entrez Gene ID
- 26255
- Gene Name
- pituitary tumor-transforming 3, pseudogene
- Gene Aliases
- PTTG3, rcPTTG1
- Chromosome Location
- Chr. 8: 66767397 - 66768005 on Build GRCh38
- UniGene ID
- Hs.647156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002734.2 |
|
1 |
563 |
|
GenBank mRNA |
AF095289.1 |
- |
1 |
534 |
|
View Full Product Details