Gene Transcripts

Gene Symbol
FOXL1
Entrez Gene ID
2300
Gene Name
forkhead box L1
Gene Aliases
FKH6, FKHL11, FREAC7
Chromosome Location
Chr. 16: 86578509 - 86581698 on Build GRCh38
UniGene ID
Hs.533830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005250.2 NP_005241.1 1 999
GenBank mRNA BC100027.1 - 1 999
BC117226.1 - 1 827 40125777

View Full Product Details