Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138689.2 | NP_619634.1 | 1, 2 | 534 | |
GenBank mRNA | AF318364.1 | - | 1 | 535 | |
BC014522.2 | - | 1, 2 | 386 | 3831227 | |
BC071884.1 | - | 1 | 289 | ||
BC094817.1 | - | 1, 2 | 417 | 5466876 | |
BT009914.1 | - | 1, 2 | 266 | ||
X91195.1 | - | 1, 2 | 518 |