Gene Transcripts
- Gene Symbol
- MPRIP
- Entrez Gene ID
- 23164
- Gene Name
- myosin phosphatase Rho interacting protein
- Gene Aliases
- M-RIP, MRIP, RHOIP3, RIP3, p116Rip
- Chromosome Location
- Chr. 17: 17042438 - 17192648 on Build GRCh38
- UniGene ID
- Hs.462341
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015134.3 |
NP_055949.2
|
7, 8 |
996 |
|
NM_201274.3 |
NP_958431.2
|
7, 8 |
996 |
|
XM_005256563.4 |
XP_005256620.1
|
7, 8 |
1301 |
|
XM_005256564.4 |
XP_005256621.3
|
7, 8 |
1301 |
|
XM_011523761.2 |
XP_011522063.1
|
7, 8 |
1672 |
|
XM_011523762.2 |
XP_011522064.1
|
7, 8 |
1655 |
|
XM_011523763.2 |
XP_011522065.1
|
7, 8 |
1655 |
|
XM_011523764.2 |
XP_011522066.1
|
7, 8 |
1655 |
|
XM_011523765.2 |
XP_011522067.1
|
7, 8 |
1655 |
|
XM_011523766.2 |
XP_011522068.2
|
7, 8 |
1655 |
|
XM_011523767.2 |
XP_011522069.2
|
7, 8 |
1655 |
|
XM_017024393.1 |
XP_016879882.1
|
7, 8 |
1655 |
|
GenBank mRNA |
AB098507.1 |
- |
7, 8 |
960 |
|
AB189741.1 |
- |
7, 8 |
901 |
|
AK092287.1 |
- |
2, 3 |
374 |
|
AL834513.1 |
- |
6, 7 |
793 |
|
AY296247.1 |
- |
7, 8 |
904 |
|
BC009982.2 |
- |
3, 4 |
433 |
|
BC075847.1 |
- |
2, 3 |
330 |
|
BC136479.1 |
- |
7, 8 |
984 |
9020468 |
View Full Product Details