Gene Transcripts

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
Chromosome Location
Chr. 1: 205504556 - 205532793 on Build GRCh38
UniGene ID
Hs.445402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002596.3 NP_002587.2 16 2812
NM_212502.2 NP_997667.1 16 2730
NM_212503.2 NP_997668.1 16 2820
XM_011509602.2 XP_011507904.1 16 2870
XM_017001423.1 XP_016856912.1 16 2965
GenBank mRNA AB209307.1 - 14 3392
AK126879.1 - 11 3603
AL161977.1 - 7 2722
BC000281.2 - 10 1897
BC011526.1 - 16 2665 3855304
BC040529.1 - 7 2818
BM695474.1 - 1 119

View Full Product Details