Gene Transcripts

Gene Symbol
TARP
Entrez Gene ID
445347
Gene Name
TCR gamma alternate reading frame protein
Gene Aliases
CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
Chromosome Location
Chr. 7: 38239577 - 38276741 on Build GRCh38
UniGene ID
Hs.534032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003799.1 NP_001003799.1 2 294
NM_001003806.1 NP_001003806.1 2 294
XM_011515407.2 XP_011513709.1 3 514
XM_017012226.1 XP_016867715.1 1 71
XM_017012227.1 XP_016867716.1 2 504
XM_017012229.1 XP_016867718.1 2 716
XM_017012230.1 XP_016867719.1 2 320
GenBank mRNA AF151103.1 - 2 294
BC030554.1 - 4 978
BC105589.1 - 3 512 4272944
BF674593.1 - 2 284
KC170727.1 - 5 651
M16804.1 - 3 573
M17325.1 - 1 213
M27334.1 - 1 234
X72500.1 - 4 660

Gene Symbol
TRGV9
Entrez Gene ID
6983
Gene Name
T cell receptor gamma variable 9
Gene Aliases
TCRGV9, TRGC1, V2
Chromosome Location
Chr. 7: 38317017 - 38317492 on Build GRCh38
UniGene ID
Hs.534032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC062761.1 - 2 289
BC072387.1 - 3 611
BC072396.1 - 5 1037
M16768.1 - 4 797

View Full Product Details