Gene Transcripts
- Gene Symbol
- CRISP2
- Entrez Gene ID
- 7180
- Gene Name
- cysteine rich secretory protein 2
- Gene Aliases
- CRISP-2, CT36, GAPDL5, TPX1, TSP1
- Chromosome Location
- Chr. 6: 49676665 - 49714198 on Build GRCh38
- UniGene ID
- Hs.2042
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001142407.2 |
NP_001135879.1
|
8, 9 |
806 |
|
NM_001142408.2 |
NP_001135880.1
|
9, 10 |
863 |
|
NM_001142417.2 |
NP_001135889.1
|
9, 10 |
843 |
|
NM_001142435.2 |
NP_001135907.1
|
8, 9 |
826 |
|
NM_001261822.1 |
NP_001248751.1
|
8, 9 |
832 |
|
NM_003296.3 |
NP_003287.1
|
9, 10 |
869 |
|
XM_005249349.1 |
XP_005249406.1
|
10, 11 |
948 |
|
XM_005249350.1 |
XP_005249407.1
|
9, 10 |
906 |
|
XM_005249351.3 |
XP_005249408.1
|
9, 10 |
926 |
|
XM_005249352.3 |
XP_005249409.1
|
9, 10 |
917 |
|
XM_005249353.3 |
XP_005249410.1
|
10, 11 |
953 |
|
XM_005249356.1 |
XP_005249413.1
|
10, 11 |
951 |
|
XM_011514841.1 |
XP_011513143.1
|
10, 11 |
1283 |
|
XM_011514842.1 |
XP_011513144.1
|
9, 10 |
1246 |
|
GenBank mRNA |
AK223337.1 |
- |
9, 10 |
806 |
|
AK292385.1 |
- |
9, 10 |
847 |
|
AY289796.1 |
- |
9, 10 |
813 |
|
AY292862.1 |
- |
9, 10 |
908 |
|
AY292863.1 |
- |
8, 9 |
807 |
|
AY773479.1 |
- |
9, 10 |
849 |
|
BC022011.2 |
- |
9, 10 |
816 |
4826427 |
BC107707.1 |
- |
8, 9 |
763 |
6618679 |
BI831679.1 |
- |
3, 4 |
218 |
|
BM552843.1 |
- |
8, 9 |
741 |
|
BP369727.1 |
- |
3, 4 |
320 |
|
M25532.1 |
- |
9, 10 |
792 |
|
X95239.1 |
- |
9, 10 |
843 |
|
View Full Product Details