Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303472.1 | NP_001290401.1 | 3, 4 | 779 | |
NM_022096.5 | NP_071379.3 | 3, 4 | 797 | ||
NM_198798.2 | NP_942093.1 | 2, 3 | 733 | ||
GenBank mRNA | AK097689.1 | - | 2, 3 | 666 | |
BC022878.2 | - | 3, 4 | 707 | 4821619 |