Gene Transcripts
- Gene Symbol
- FNDC3B
- Entrez Gene ID
- 64778
- Gene Name
- fibronectin type III domain containing 3B
- Gene Aliases
- FAD104, PRO4979, YVTM2421
- Chromosome Location
- Chr. 3: 172039628 - 172400703 on Build GRCh38
- UniGene ID
- Hs.744888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135095.1 |
NP_001128567.1
|
3, 4 |
294 |
|
NM_022763.3 |
NP_073600.3
|
3, 4 |
367 |
|
XM_017007062.1 |
XP_016862551.1
|
3, 4 |
347 |
|
XM_017007064.1 |
XP_016862553.1
|
3, 4 |
347 |
|
GenBank mRNA |
AB098597.1 |
- |
2, 3 |
252 |
|
AK127826.1 |
- |
3, 4 |
347 |
|
AK223599.1 |
- |
3, 4 |
398 |
|
AK314478.1 |
- |
3, 4 |
294 |
|
AY358367.1 |
- |
3, 4 |
336 |
|
BC039297.1 |
- |
3, 4 |
258 |
5297602 |
BX648340.1 |
- |
3, 4 |
289 |
|
View Full Product Details