Gene Transcripts

Gene Symbol
SZT2
Entrez Gene ID
23334
Gene Name
seizure threshold 2 homolog (mouse)
Gene Aliases
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B
Chromosome Location
Chr. 1: 43389884 - 43454247 on Build GRCh38
UniGene ID
Hs.643560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015284.3 NP_056099.3 71 10347
XM_005270686.3 XP_005270743.1 73 10579
XM_006710501.3 XP_006710564.1 72 10466
XM_011541106.2 XP_011539408.1 72 10617
XM_011541107.2 XP_011539409.1 69 10114
XM_017000819.1 XP_016856308.1 73 10426
XM_017000820.1 XP_016856309.1 72 10408
XM_017000821.1 XP_016856310.1 69 10100
GenBank mRNA AB007936.2 - 57 8199
AK001249.2 - 6 1158
AK027078.1 - 12 1892
AK126972.1 - 10 2249
AL117402.1 - 6 1141
BC041069.1 - 13 2028
BC052802.1 - 14 2042
BC082968.1 - 14 2047
BC151232.1 - 57 8199 8860124

View Full Product Details