Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
79785
Gene Name
RERG like
Gene Aliases
Chromosome Location
Chr. 12: 18080869 - 18090193 on Build GRCh38
UniGene ID
Hs.732831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286201.1 NP_001273130.1 5 663
NM_024730.3 NP_079006.1 6 820
NR_104413.1 5 560
GenBank mRNA AK026308.1 - 6 746
BC042888.1 - 6 807 5198817
BX422875.2 - 5 611

View Full Product Details