Gene Transcripts

Gene Symbol
TMC7
Entrez Gene ID
79905
Gene Name
transmembrane channel like 7
Gene Aliases
Chromosome Location
Chr. 16: 18983934 - 19063942 on Build GRCh38
UniGene ID
Hs.187377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160364.2 NP_001153836.1 16 3373
NM_001324263.1 NP_001311192.1 16 3262
NM_001324265.1 NP_001311194.1 15 3119
NM_001324268.1 NP_001311197.1 15 3127
NM_024847.4 NP_079123.3 16 3275
NR_136733.1 15 3106
GenBank mRNA AK024893.1 - 12 2484
AY236498.1 - 16 3209
AY263165.1 - 16 3204
BC036205.1 - 16 3204 5261157
CR933669.1 - 16 3372

Gene Symbol
LOC102723385
Entrez Gene ID
102723385
Gene Name
uncharacterized LOC102723385
Gene Aliases
Chromosome Location
Chr. 16: 19062754 - 19067691 on Build GRCh38
UniGene ID
Hs.569608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AW663894.1 - 1 64

View Full Product Details