Gene Transcripts
- Gene Symbol
- THOC7
- Entrez Gene ID
- 80145
- Gene Name
- THO complex 7
- Gene Aliases
- NIF3L1BP1, fSAP24, hTREX30
- Chromosome Location
- Chr. 3: 63833870 - 63864478 on Build GRCh38
- UniGene ID
- Hs.288151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285404.1 |
NP_001272333.1
|
2, 3 |
407 |
|
NM_025075.3 |
NP_079351.2
|
2, 3 |
276 |
|
XM_006713339.3 |
XP_006713402.1
|
2, 3 |
225 |
|
GenBank mRNA |
AI391685.1 |
- |
2, 3 |
256 |
|
AK027098.1 |
- |
2, 3 |
186 |
|
AK225110.1 |
- |
2, 3 |
164 |
|
BC020599.2 |
- |
1, 2 |
124 |
|
BC065012.1 |
- |
2, 3 |
146 |
6167514 |
BI914906.1 |
- |
2, 3 |
407 |
|
CR457373.1 |
- |
2, 3 |
143 |
|
HY023143.1 |
- |
2, 3 |
276 |
|
View Full Product Details