Gene Transcripts

Gene Symbol
ZFP2
Entrez Gene ID
80108
Gene Name
ZFP2 zinc finger protein
Gene Aliases
ZNF751, zfp-2
Chromosome Location
Chr. 5: 178895894 - 178933212 on Build GRCh38
UniGene ID
Hs.654533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030613.3 NP_085116.2 5 2328
GenBank mRNA AK025281.1 - 1 1643
AK131367.1 - 5 2307
BC142989.1 - 5 2297 8860436

View Full Product Details