Gene Transcripts

Gene Symbol
SNX27
Entrez Gene ID
81609
Gene Name
sorting nexin family member 27
Gene Aliases
MRT1, MY014
Chromosome Location
Chr. 1: 151612027 - 151699083 on Build GRCh38
UniGene ID
Hs.192326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030918.5 NP_112180.4 11, 12 1602
XM_005245511.4 XP_005245568.1 11, 12 1630
XM_017002417.1 XP_016857906.1 10, 11 1303
GenBank mRNA AB007957.2 - 11, 12 1571
AF060509.1 - 5, 6 543
BC012184.1 - 11, 12 1592
BC051817.1 - 8, 9 851
BC100998.1 - 11, 12 1602 40006277
BC100999.1 - 11, 12 1600 40006279
BC101000.1 - 11, 12 1602 40006281
BC101822.1 - 11, 12 1637 8069328
BC101824.1 - 11, 12 1637 8069330

View Full Product Details