Gene Transcripts

Gene Symbol
CCDC70
Entrez Gene ID
83446
Gene Name
coiled-coil domain containing 70
Gene Aliases
Chromosome Location
Chr. 13: 51861981 - 51866236 on Build GRCh38
UniGene ID
Hs.120573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031290.2 NP_112580.2 2 1144
GenBank mRNA AL136887.1 - 2 1115

View Full Product Details