Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300948.1 | NP_001287877.1 | 6 | 1199 | |
NM_031490.3 | NP_113678.2 | 7 | 1331 | ||
XM_017023755.1 | XP_016879244.1 | 7 | 1198 | ||
XM_017023756.1 | XP_016879245.1 | 6 | 1064 | ||
GenBank mRNA | AJ548761.1 | - | 7 | 1198 | |
AK027544.1 | - | 7 | 1164 | ||
AK027666.1 | - | 3 | 366 | ||
AK074775.1 | - | 7 | 1196 | ||
AK127125.1 | - | 7 | 963 | ||
AK294779.1 | - | 6 | 1011 | ||
AL834201.1 | - | 6 | 697 | ||
BC093910.1 | - | 7 | 1165 | 7939755 | |
BC093912.1 | - | 7 | 1165 | 7939757 | |
BC110434.1 | - | 3 | 347 | ||
BC143246.1 | - | 6 | 1033 | 9051747 |