Gene Transcripts

Gene Symbol
CUX1
Entrez Gene ID
1523
Gene Name
cut like homeobox 1
Gene Aliases
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
Chromosome Location
Chr. 7: 101815904 - 102283958 on Build GRCh38
UniGene ID
Hs.191482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202543.1 NP_001189472.1 2 240
NM_001202544.2 NP_001189473.1 2 240
NM_001202545.2 NP_001189474.1 2 240
NM_001913.4 NP_001904.2 2 240
NM_181500.3 NP_852477.1 2 240
NM_181552.3 NP_853530.2 2 118
XM_005250150.2 XP_005250207.1 2 639
XM_005250151.2 XP_005250208.1 2 642
XM_005250154.3 XP_005250211.1 2 496
XM_006715854.2 XP_006715917.1 2 639
XM_006715855.1 XP_006715918.1 2 496
XM_011515823.2 XP_011514125.1 2 637
XM_011515825.1 XP_011514127.1 2 205
XM_017011759.1 XP_016867248.1 2 205
XM_017011760.1 XP_016867249.1 2 205
GenBank mRNA AK122726.1 - 2 123
AK125097.1 - 2 135
AK222832.1 - 2 136
AK297548.1 - 2 126
BC025422.2 - 2 121
BC066592.1 - 2 133 5740343
DC410774.1 - 2 240
L12579.1 - 2 132
M74099.1 - 2 123

View Full Product Details