Gene Transcripts
- Gene Symbol
- PRPS2
- Entrez Gene ID
- 5634
- Gene Name
- phosphoribosyl pyrophosphate synthetase 2
- Gene Aliases
- PRSII
- Chromosome Location
- Chr. X: 12791355 - 12824227 on Build GRCh38
- UniGene ID
- Hs.654581
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039091.2 |
NP_001034180.1
|
1, 2 |
265 |
|
NM_002765.4 |
NP_002756.1
|
1, 2 |
265 |
|
GenBank mRNA |
AK313246.1 |
- |
1, 2 |
237 |
|
BC030019.2 |
- |
1, 2 |
206 |
|
BC040483.1 |
- |
1, 2 |
208 |
5272983 |
BC110875.1 |
- |
1, 2 |
199 |
6472836 |
BC119662.1 |
- |
1, 2 |
197 |
40116022 |
BC119663.1 |
- |
1, 2 |
197 |
40116023 |
CR988262.1 |
- |
1, 2 |
214 |
|
DA478574.1 |
- |
1, 2 |
265 |
|
Y00971.1 |
- |
1, 2 |
182 |
|
View Full Product Details