Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005189.2 | NP_005180.1 | 4 | 308 | |
NM_032647.3 | NP_116036.1 | 4 | 308 | ||
XM_011525382.1 | XP_011523684.1 | 4 | 348 | ||
XM_011525383.2 | XP_011523685.1 | 4 | 1310 | ||
GenBank mRNA | BC004252.1 | - | 4 | 292 | |
BC119759.1 | - | 4 | 279 | ||
BC119760.1 | - | 4 | 279 | ||
CN367085.1 | - | 4 | 309 | ||
CN367087.1 | - | 4 | 232 | ||
CV815254.1 | - | 4 | 209 |