Gene Transcripts

Gene Symbol
FOXE3
Entrez Gene ID
2301
Gene Name
forkhead box E3
Gene Aliases
FKHL12, FREAC8
Chromosome Location
Chr. 1: 47416072 - 47418052 on Build GRCh38
UniGene ID
Hs.112968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012186.2 NP_036318.1 1 1822
GenBank mRNA AF275722.1 - 1 1833

View Full Product Details