Gene Transcripts

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
Chromosome Location
Chr. 1: 11654375 - 11663327 on Build GRCh38
UniGene ID
Hs.556006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014765.1 NP_001014765.1 7 1469
NM_001304790.1 NP_001291719.1 6 1002
NM_001304791.1 NP_001291720.1 6 1208
NM_033182.5 NP_149438.2 6 1097
NM_183412.2 NP_904319.1 5 1083
NM_183413.2 NP_904320.1 5 972
NR_130909.1 5 838
XM_005263535.1 XP_005263592.1 6 1039
XM_005263536.4 XP_005263593.1 7 1433
XM_005263537.1 XP_005263594.1 6 1179
XM_011542435.1 XP_011540737.1 7 1501
XM_017002842.1 XP_016858331.1 7 950
XM_017002843.1 XP_016858332.1 7 981
GenBank mRNA AK055344.1 - 5 1083
AK296025.1 - 5 818
AK301418.1 - 6 1077
AK316284.1 - 5 1216
AY040878.1 - 6 1053
BC007832.2 - 5 919 4054414
CR749368.1 - 7 1458

View Full Product Details