Gene Transcripts

Gene Symbol
LACRT
Entrez Gene ID
90070
Gene Name
lacritin
Gene Aliases
Chromosome Location
Chr. 12: 54630839 - 54634879 on Build GRCh38
UniGene ID
Hs.307096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033277.1 NP_150593.1 4 380
GenBank mRNA AF238867.1 - 4 394
BC062217.1 - 4 400 30327037
BC069317.1 - 4 380 7262178

View Full Product Details