Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267623.1 | NP_001254552.1 | 6 | 635 | |
NM_001324236.1 | NP_001311165.1 | 7 | 761 | ||
NM_001324237.1 | NP_001311166.1 | 8 | 666 | ||
NM_001324238.1 | NP_001311167.1 | 7 | 669 | ||
NM_033453.3 | NP_258412.1 | 8 | 758 | ||
NM_181493.3 | NP_852470.1 | 8 | 707 | ||
NR_052000.1 | 7 | 601 | |||
NR_052001.2 | 7 | 679 | |||
NR_052002.1 | 7 | 693 | |||
XM_006723564.3 | XP_006723627.1 | 9 | 1088 | ||
XM_006723565.3 | XP_006723628.1 | 7 | 965 | ||
GenBank mRNA | AF026816.2 | - | 8 | 617 | |
AF219116.1 | - | 8 | 670 | ||
AF334680.1 | - | 7 | 550 | ||
BC010138.2 | - | 8 | 602 | 4124624 | |
BE275997.1 | - | 7 | 635 | ||
BF969192.1 | - | 7 | 600 | ||
BM560812.1 | - | 760 | |||
BQ049090.1 | - | 7 | 540 | ||
EF199841.1 | - | 8 | 605 | ||
EF213026.1 | - | 8 | 554 | ||
KU178148.1 | - | 8 | 566 | ||
KU178149.1 | - | 6 | 443 |