Gene Transcripts

Gene Symbol
WDR89
Entrez Gene ID
112840
Gene Name
WD repeat domain 89
Gene Aliases
C14orf150, MSTP050
Chromosome Location
Chr. 14: 63597039 - 63641923 on Build GRCh38
UniGene ID
Hs.655666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008726.2 NP_001008726.1 4 486
NM_001258272.1 NP_001245201.1 3 882
NM_080666.3 NP_542397.1 3 377
XM_011536383.2 XP_011534685.1 2 416
XM_011536385.2 XP_011534687.1 4 384
GenBank mRNA AF115513.1 - 3 309
AK095324.1 - 3 325
BC010698.1 - 2 269 3870073
BI603622.1 - 3 333
BP278550.1 - 4 397
BX248008.1 - 3 366
DA882425.1 - 3 378

View Full Product Details