Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008726.2 | NP_001008726.1 | 4 | 1525 | |
NM_001258272.1 | NP_001245201.1 | 3 | 1921 | ||
NM_080666.3 | NP_542397.1 | 3 | 1416 | ||
XM_011536383.2 | XP_011534685.1 | 2 | 1455 | ||
XM_011536385.2 | XP_011534687.1 | 4 | 1423 | ||
GenBank mRNA | AF115513.1 | - | 3 | 1348 | |
AK095324.1 | - | 3 | 1364 | ||
BC010698.1 | - | 2 | 1308 | 3870073 | |
BX248008.1 | - | 3 | 1405 |