Gene Transcripts
- Gene Symbol
- CLEC4C
- Entrez Gene ID
- 170482
- Gene Name
- C-type lectin domain family 4 member C
- Gene Aliases
- BDCA2, CD303, CLECSF11, CLECSF7, DLEC, HECL, PRO34150
- Chromosome Location
- Chr. 12: 7729245 - 7749473 on Build GRCh38
- UniGene ID
- Hs.351812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_130441.2 |
NP_569708.1
|
4, 5 |
719 |
|
NM_203503.1 |
NP_987099.1
|
3, 4 |
626 |
|
XM_017018940.1 |
XP_016874429.1
|
3, 4 |
358 |
|
GenBank mRNA |
AF293615.1 |
- |
4, 5 |
719 |
|
AF325459.1 |
- |
3, 4 |
254 |
|
AF325460.1 |
- |
2, 3 |
142 |
|
AK122596.1 |
- |
3, 4 |
402 |
|
AY358223.1 |
- |
3, 4 |
385 |
|
BC074967.2 |
- |
3, 4 |
272 |
30915289 |
BC074968.2 |
- |
3, 4 |
272 |
30915508 |
BC102015.1 |
- |
3, 4 |
329 |
40030336 |
BC102016.1 |
- |
3, 4 |
329 |
40030338 |
BC102017.1 |
- |
3, 4 |
328 |
40030339 |
BC114338.1 |
- |
3, 4 |
329 |
40030323 |
View Full Product Details