Gene Transcripts
- Gene Symbol
- RHBDL3
- Entrez Gene ID
- 162494
- Gene Name
- rhomboid like 3
- Gene Aliases
- RHBDL4, VRHO
- Chromosome Location
- Chr. 17: 32266167 - 32324661 on Build GRCh38
- UniGene ID
- Hs.655027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138328.2 |
NP_612201.1
|
3, 4 |
301 |
|
XM_006721733.3 |
XP_006721796.1
|
2, 3 |
278 |
|
XM_006721734.3 |
XP_006721797.1
|
2, 3 |
670 |
|
XM_017024272.1 |
XP_016879761.1
|
3, 4 |
306 |
|
XM_017024273.1 |
XP_016879762.1
|
2, 3 |
281 |
|
XM_017024274.1 |
XP_016879763.1
|
2, 3 |
609 |
|
GenBank mRNA |
AJ313480.1 |
- |
3, 4 |
301 |
|
BC100975.1 |
- |
3, 4 |
300 |
40005244 |
BC100976.1 |
- |
3, 4 |
301 |
|
BC100977.1 |
- |
2, 3 |
277 |
40005246 |
View Full Product Details