Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138409.2 | NP_612418.2 | 4 | 1508 | |
XM_011535400.2 | XP_011533702.1 | 4 | 1434 | ||
XM_017010219.1 | XP_016865708.1 | 5 | 1806 | ||
XM_017010220.1 | XP_016865709.1 | 4 | 2075 | ||
GenBank mRNA | AJ420596.1 | - | 1 | 747 | |
AK090775.1 | - | 4 | 1508 | ||
AK307078.1 | - | 3 | 1336 | ||
BC010003.2 | - | 4 | 1453 | 4134852 | |
BC039855.1 | - | 4 | 1372 | 3850270 |